(S)-2-((tert-Butoxycarbonyl)amino)-4-phenylbutanoic acid ≥98.0%

Supplier: TCI America

Synonyms: N-Boc-L-homophenylalanine, (S)-2-(tert-Butoxycarbonylamino)-4-phenylbutyric Acid, Boc-Homophe-OH, (S)-2-(Boc-amino)-4-phenylbutyric Acid

B4356-1G B4356-5G
TCB4356-1GEA 26.86 CAD
TCB4356-1G TCB4356-5G
(S)-2-((tert-Butoxycarbonyl)amino)-4-phenylbutanoic acid ≥98.0%
(S)-2-((tert-Butoxycarbonyl)amino)-4-phenylbutanoic acid

CAS Number: 100564-78-1
MDL Number: MFCD00076904
Molecular Formula: C15H21NO4
Molecular Weight: 279.34
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 78
Specific rotation [a]20/D: 6 deg (C=1, EtOH)
Storage Temperature: 0-10°C


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-L-homophenylalanine, (S)-2-(tert-Butoxycarbonylamino)-4-phenylbutyric Acid, (S)-2-(Boc-amino)-4-phenylbutyric Acid

Formula: C₁₅H₂₁NO₄
MW: 279.34 g/mol
Melting Pt: 78 °C
MDL Number: MFCD00076904
CAS Number: 100564-78-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eddd3c35f64fc4b2978a3f77341b02a62f163a4f&vid=eddd3c35f64fc4b2978a3f77341b02a62f163a4f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR