tert-Butyl 4-((methylsulfonyl)oxy)piperidine-1-carboxylate ≥98.0%

Supplier: TCI America

Synonyms: tert-Butyl 4-(methanesulfonyloxy)piperidine-1-carboxylate, 1-Boc-4-(methanesulphonyloxy)piperidine, tert-Butyl 4-((methylsulphonyl)oxy)piperidine-1-carboxylate, 1-Boc-4-(methanesulfonyloxy)piperidine, 4-(Methanesulfonyloxy)piperidine-1-carboxylic acid tert-Butyl ester, 1-(tert-Butoxycarbonyl)-4-piperidyl Methanesulfonate, tert-Butyl 4-(Methanesulphonyloxy)piperidine-1-carboxylate, 4-(Methanesulphonyloxy)piperidine-1-carboxylic acid tert-Butyl ester, 1-(tert-Butoxycarbonyl)-4-(methanesulphonyloxy)piperidine, tert-Butyl 4-((methylsulfonyl)oxy)piperidine-1-carboxylate, 1-(tert-Butoxycarbonyl)-4-piperidyl Methanesulphonate, 1-(tert-Butoxycarbonyl)-4-(methanesulfonyloxy)piperidine

B4353-25G B4353-5G
TCB4353-25GEA 417.36 CAD
TCB4353-25G TCB4353-5G
tert-Butyl 4-((methylsulfonyl)oxy)piperidine-1-carboxylate ≥98.0%
tert-Butyl 4-((methylsulfonyl)oxy)piperidine-1-carboxylate

CAS Number: 141699-59-4
MDL Number: MFCD04116220
Molecular Formula: C11H21NO5S
Molecular Weight: 279.35
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 95


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: 1-(tert-Butoxycarbonyl)-4-piperidyl Methanesulfonate, 1-Boc-4-(methanesulfonyloxy)piperidine, tert-Butyl 4-(Methanesulfonyloxy)piperidine-1-carboxylate, 4-(Methanesulfonyloxy)piperidine-1-carboxylic Acid tert-Butyl Ester

Formula: C₁₁H₂₁NO₅S
Melting Pt: 95 °C
MDL Number: MFCD04116220
CAS Number: 141699-59-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2a03d89ab290e0ee4577ad1bf411e316dcc52de&vid=e2a03d89ab290e0ee4577ad1bf411e316dcc52de&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR