2-Bromopyrazine ≥98.0% (by GC)

Supplier: TCI America

B4352-1G B4352-5G
TCB4352-1GEA 158 CAD
TCB4352-1G TCB4352-5G
2-Bromopyrazine ≥98.0% (by GC)
2-Bromopyrazine

CAS Number: 56423-63-3
MDL Number: MFCD08275680
Molecular Formula: C4H3BrN2
Molecular Weight: 158.99
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 61
Flash Point (°C): 92
Specific Gravity (20/20): 1.73
Storage Temperature: 0-10°C

Formula: C₄H₃BrN₂
MW: 158.9800 g/mol
Boiling Pt: 61 °C
Density: 1.73 (20 °C)
Flash Pt: 92 °C
MDL Number: MFCD08275680
CAS Number: 56423-63-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b4196e9244e334eed5eda7afd12dc3f146f1eb2&vid=2b4196e9244e334eed5eda7afd12dc3f146f1eb2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR