Bergenin ≥98.0%

Supplier: TCI America

Synonyms: (2R,3S,4S,4aR,10bS)-3,4,8,10-Tetrahydroxy-2-(hydroxymethyl)-9-methoxy-3,4,4a,10b-tetrahydropyrano[3,2-c]isochromen-6(2H)-one

B4349-1G B4349-5G
TCB4349-1GEA 274.92 CAD
TCB4349-1G TCB4349-5G
Bergenin ≥98.0%
Bergenin

CAS Number: 477-90-7
MDL Number: MFCD00133120
Molecular Formula: C14H16O9
Molecular Weight: 328.27
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 240
Specific rotation [a]20/D: -39 deg (C=2, MeOH)
Lambda max.: 277 nm (EtOH)Storage Temperature: 0-10°C

Formula: C₁₄H₁₆O₉
MW: 328.27 g/mol
Melting Pt: 240 °C
MDL Number: MFCD00133120
CAS Number: 477-90-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a75554edd6e844ca389e47016f38d775aa51502&vid=4a75554edd6e844ca389e47016f38d775aa51502&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR