3-Bromo-5-iodopyridine ≥98.0% (by GC)

Supplier: TCI America

B4346-1G B4346-5G
TCB4346-1GEA 197.99 CAD
TCB4346-1G TCB4346-5G
3-Bromo-5-iodopyridine ≥98.0% (by GC)
3-Bromo-5-iodopyridine

CAS Number: 233770-01-9
MDL Number: MFCD03086019
Molecular Formula: C5H3BrIN
Molecular Weight: 283.89
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 133

Formula: C₅H₃BrIN
MW: 283.89 g/mol
Melting Pt: 133 °C
MDL Number: MFCD03086019
CAS Number: 233770-01-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8063de0046b81b30598f02f71c6d7a93a2084a0&vid=e8063de0046b81b30598f02f71c6d7a93a2084a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR