4-Bromodibenzofuran ≥98.0%

Supplier: TCI America

Synonyms: 4-Bromodibenzo[b,d]furan

B4345-1G B4345-5G
TCB4345-1GEA 128.93 CAD
TCB4345-1G TCB4345-5G
4-Bromodibenzofuran ≥98.0%
4-Bromodibenzofuran

CAS Number: 89827-45-2
MDL Number: MFCD00185685
Molecular Formula: C12H7BrO
Molecular Weight: 247.09
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 70

Formula: C₁₂H₇BrO
MW: 247.09 g/mol
Melting Pt: 70 °C
MDL Number: MFCD00185685
CAS Number: 89827-45-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=439f35184ee2857287899154845363ae749f4d9d&vid=439f35184ee2857287899154845363ae749f4d9d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR