2,4-Bis[4-(diethylamino)-2-hydroxyphenyl]squaraine ≥98.0%
Supplier: TCI America
B4342-1G
B4342-5G
TCB4342-1GEA
120.08
CAD
TCB4342-1G
TCB4342-5G
2,4-Bis[4-(diethylamino)-2-hydroxyphenyl]squaraine ≥98.0%
2,4-Bis[4-(diethylamino)-2-hydroxyphenyl]squaraine
CAS Number: 68842-66-0
MDL Number: MFCD00142492
Molecular Formula: C24H28N2O4
Molecular Weight: 408.50
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Lambda max.: 648 nm (CH2Cl2)
Formula:
C₂₄H₂₈N₂O₄ MW: 408.50 g/mol |
MDL Number:
MFCD00142492 CAS Number: 68842-66-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...