2-Benzylbenzimidazole hydrochloride ≥98.0% (by HPLC)

Supplier: TCI America

B4341-25G B4341-5G
TCB4341-25GEA 409.85 CAD
TCB4341-25G TCB4341-5G
2-Benzylbenzimidazole hydrochloride ≥98.0% (by HPLC)
2-Benzylbenzimidazole hydrochloride

CAS Number: 1212-48-2
MDL Number: MFCD00443659
Molecular Formula: C14H12N2
Molecular Weight: 244.72
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal

Formula: C₁₄H₁₂N₂
MW: 244.72 g/mol
MDL Number: MFCD00443659
CAS Number: 1212-48-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97dcb29f0e85ac44d41284b19f130b049a00e9a1&vid=97dcb29f0e85ac44d41284b19f130b049a00e9a1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR