trans-4-Bromocinnamaldehyde ≥98.0% (by GC)
Supplier: TCI America
B4337-1G
B4337-5G
TCB4337-1GEA
93.22
CAD
TCB4337-1G
TCB4337-5G
trans-4-Bromocinnamaldehyde ≥98.0% (by GC)
trans-4-Bromocinnamaldehyde
CAS Number: 49678-04-8
MDL Number: MFCD03032509
Molecular Formula: C9H7BrO
Molecular Weight: 211.06
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 80
Lambda max.: 298 nm (CHCl3)
Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: trans-3-(4-Bromophenyl)acrylaldehyde, trans-3-(4-Bromophenyl)-2-propenal, trans-3-(4-Bromophenyl)acrolein
Formula:
C₉H₇BrO Melting Pt: 80 °C |
MDL Number:
MFCD03032509 CAS Number: 49678-04-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...