(4-Boc-aminophenyl)boronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
(4-Boc-Aminophenyl)Boronic Acid, 4-Boc-aminophenylboronic acid, (4-Boc-Aminophenyl)boronic acid, 4-(N-BOC-amino)phenylboronic acid, [4-[(2-methylpropan-2-yl)oxycarbonylamino]phenyl]boronic Acid
CAS Number: 380430-49-9
MDL Number: MFCD02093054
Molecular Formula: C11H16BNO4
Molecular Weight: 237.06
Form: Crystal
Color: White
Melting point (°C): 204
Other Product Names / Synonyms: 4-(Boc-amino)phenylboronic Acid, 4-[(tert-Butoxycarbonyl)amino]benzeneboronic Acid, 4-(Boc-amino)benzeneboronic Acid
Formula:
C₁₁H₁₆BNO₄ MW: 237.062 g/mol Melting Pt: 204 °C |
MDL Number:
MFCD02093054 CAS Number: 380430-49-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...