6-Bromo-1H-indole-2-carboxylic acid ≥97.0% (by GC, titration analysis)
Supplier: TCI America
B4334-1G
B4334-5G
TCB4334-1GEA
191.18
CAD
TCB4334-1G
TCB4334-5G
6-Bromo-1H-indole-2-carboxylic acid ≥97.0% (by GC, titration analysis)
6-Bromo-1H-indole-2-carboxylic acid
CAS Number: 16732-65-3
MDL Number: MFCD02664469
Molecular Formula: C9H6BrNO2
Molecular Weight: 240.06
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 224
Lambda max.: 308 nm (EtOH)
Formula:
C₉H₆BrNO₂ Melting Pt: 224 °C |
MDL Number:
MFCD02664469 CAS Number: 16732-65-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...