N-tert-Butyl-2-thiophenesulfonamide ≥98.0% (by GC)

Supplier: TCI America

B4330-1G B4330-5G
TCB4330-1GEA 28.44 CAD
TCB4330-1G TCB4330-5G
N-tert-Butyl-2-thiophenesulfonamide ≥98.0% (by GC)
N-tert-Butyl-2-thiophenesulfonamide

CAS Number: 100342-30-1
MDL Number: MFCD02047252
Molecular Formula: C8H13NO2S2
Molecular Weight: 219.32
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 84


Other Product Names / Synonyms: 2-(tert-Butylaminosulfonyl)thiophene

Formula: C₈H₁₃NO₂S₂
Melting Pt: 84 °C
MDL Number: MFCD02047252
CAS Number: 100342-30-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31ebca46703b7bc4b038ef53401ee1daac805d5d&vid=31ebca46703b7bc4b038ef53401ee1daac805d5d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR