2-Bromo-5-thiazolecarboxylic acid ≥97.0%

Supplier: TCI America

Synonyms: 2-Bromothiazole-5-carboxylic acid

B4329-1G B4329-5G
TCB4329-1GEA 79 CAD
TCB4329-1G TCB4329-5G
2-Bromo-5-thiazolecarboxylic acid ≥97.0%
2-Bromo-5-thiazolecarboxylic acid

CAS Number: 54045-76-0
MDL Number: MFCD04115730
Molecular Formula: C4H2BrNO2S
Molecular Weight: 208.03
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 175

Formula: C₄H₂BrNO₂S
Melting Pt: 175 °C
MDL Number: MFCD04115730
CAS Number: 54045-76-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d05c647ed481e3c8098791a1614b1ab3b0a05d17&vid=d05c647ed481e3c8098791a1614b1ab3b0a05d17&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR