6-Bromo-4-quinazolone ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B4328-25G B4328-5G
TCB4328-25GEA 323.9 CAD
TCB4328-25G TCB4328-5G
6-Bromo-4-quinazolone ≥98.0% (by GC, titration analysis)
6-Bromo-4-quinazolone

CAS Number: 32084-59-6
MDL Number: MFCD00115131
Molecular Formula: C8H5BrN2O
Molecular Weight: 225.05
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal


Other Product Names / Synonyms: 6-Bromo-4-quinazolinone, 6-Bromo-4-quinazolinol

Formula: C₈H₅BrN₂O
MDL Number: MFCD01219541
CAS Number: 32084-59-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0898695fbfd7871d105e818029b78a90dbbdd564&vid=d55415bd42441979ab4ef70d780e023d62448f45&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR