(5-Bromopyridin-2-yl)hydrazine ≥96.0% (by GC, titration analysis)
Supplier: TCI America
B4327-1G
B4327-5G
TCB4327-1GEA
64.78
CAD
TCB4327-1G
TCB4327-5G
(5-Bromopyridin-2-yl)hydrazine ≥96.0% (by GC, titration analysis)
(5-Bromopyridin-2-yl)hydrazine
CAS Number: 77992-44-0
MDL Number: MFCD00513424
Molecular Formula: C5H6BrN3
Molecular Weight: 188.03
Purity/Analysis Method: >96.0% (GC,T)
Form: Crystal
Melting point (°C): 134
Other Product Names / Synonyms: (5-Bromo-2-pyridyl)hydrazine
Formula:
C₅H₆BrN₃ Melting Pt: 134 °C |
MDL Number:
MFCD00513424 CAS Number: 77992-44-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...