2-Bromo-9,10-bis(2-naphthalenyl)anthracene ≥98.0% (by HPLC)
Supplier: TCI America
B4321-1G
B4321-5G
TCB4321-1GEA
88.48
CAD
TCB4321-1G
TCB4321-5G
2-Bromo-9,10-bis(2-naphthalenyl)anthracene ≥98.0% (by HPLC)
2-Bromo-9,10-bis(2-naphthalenyl)anthracene
CAS Number: 474688-76-1
MDL Number: MFCD11045064
Molecular Formula: C34H21Br
Molecular Weight: 509.45
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Formula:
C34H21Br |
MDL Number:
MFCD11045064 CAS Number: 474688-76-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...