4-Bromo-3-fluorobenzaldehyde ≥98.0% (by GC)

Supplier: TCI America

B4319-25G B4319-5G
TCB4319-25GEA 423.12 CAD
TCB4319-25G TCB4319-5G
4-Bromo-3-fluorobenzaldehyde ≥98.0% (by GC)
4-Bromo-3-fluorobenzaldehyde

CAS Number: 133059-43-5
MDL Number: MFCD03095000
Molecular Formula: C7H4BrFO
Molecular Weight: 203.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 57

Formula: C₇H₄BrFO
MW: 203.01 g/mol
Melting Pt: 57 °C
MDL Number: MFCD03095000
CAS Number: 133059-43-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6f4325d10790d640d92c6a4628eb04218a45a8e&vid=b6f4325d10790d640d92c6a4628eb04218a45a8e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR