7-Benzyloxy-6-methoxy-3H-quinazolin-4-one ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B4317-1G B4317-5G
TCB4317-1GEA 109.02 CAD
TCB4317-1G TCB4317-5G
7-Benzyloxy-6-methoxy-3H-quinazolin-4-one ≥98.0% (by HPLC, titration analysis)
7-Benzyloxy-6-methoxy-3H-quinazolin-4-one

CAS Number: 179688-01-8
MDL Number: MFCD04115119
Molecular Formula: C16H14N2O3
Molecular Weight: 282.30
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 269


Other Product Names / Synonyms: 7-Benzyloxy-6-methoxy-4-quinazolinol, 7-Benzyloxy-4-hydroxy-6-methoxyquinazoline

Formula: C₁₆H₁₄N₂O₃
Melting Pt: 269 °C
MDL Number: MFCD04115119
CAS Number: 179688-01-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc84d4d38d8ec83a5c55ad6290caef9704f1d9d2&vid=dc84d4d38d8ec83a5c55ad6290caef9704f1d9d2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR