4-Bromo-2-tert-butylphenol ≥95.0% (by GC)

Supplier: TCI America

B4315-1G B4315-5G
TCB4315-1GEA 213.3 CAD
TCB4315-1G TCB4315-5G
4-Bromo-2-tert-butylphenol ≥95.0% (by GC)
4-Bromo-2-tert-butylphenol

CAS Number: 10323-39-4
MDL Number: MFCD00192645
Molecular Formula: C10H13BrO
Molecular Weight: 229.12
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.37
Storage Temperature: 0-10°C

Formula: C₁₀H₁₃BrO
Density: 1.37 (20 °C)
MDL Number: MFCD00192645
CAS Number: 10323-39-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3eecfc281a02c4f3582b51f07db2c61867d8f197&vid=3eecfc281a02c4f3582b51f07db2c61867d8f197&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR