4-tert-Butylphenylacetic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B4307-25G B4307-5G
TCB4307-25GEA 172.22 CAD
TCB4307-25G TCB4307-5G
4-tert-Butylphenylacetic acid ≥98.0% (by GC, titration analysis)
4-tert-Butylphenylacetic acid

CAS Number: 32857-63-9
MDL Number: MFCD00082593
Molecular Formula: C12H16O2
Molecular Weight: 192.26
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 81

Formula: C₁₂H₁₆O₂
Melting Pt: 81 °C
MDL Number: MFCD00082593
CAS Number: 32857-63-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5677d0956e6a2c352d586f09162f5960e9da5272&vid=5677d0956e6a2c352d586f09162f5960e9da5272&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR