4-[(tert-Butoxycarbonylamino)methyl]benzoic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
B4305-1G
B4305-5G
TCB4305-1GEA
63.2
CAD
TCB4305-1G
TCB4305-5G
4-[(tert-Butoxycarbonylamino)methyl]benzoic acid ≥98.0% (by HPLC, titration analysis)
4-[(tert-Butoxycarbonylamino)methyl]benzoic acid
CAS Number: 33233-67-9
MDL Number: MFCD00228182
Molecular Formula: C13H17NO4
Molecular Weight: 251.28
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 168
Other Product Names / Synonyms: 4-[(Boc-amino)methyl]benzoic Acid
Formula:
C₁₃H₁₇NO₄ Melting Pt: 168 °C |
MDL Number:
MFCD00228182 CAS Number: 33233-67-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...