Bis(2-ethylhexyl)isophthalate ≥98.0%

Supplier: TCI America

Synonyms: Isophthalic Acid Bis(2-ethylhexyl) Ester

B4302-25G B4302-500G
TCB4302-25GEA 64.27 CAD
TCB4302-25G TCB4302-500G
Bis(2-ethylhexyl)isophthalate ≥98.0%
Bis(2-ethylhexyl)isophthalate

CAS Number: 137-89-3
MDL Number: MFCD00152284
Molecular Formula: C24H38O4
Molecular Weight: 390.56
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 211
Flash Point (°C): 233
Specific Gravity (20/20): 0.98


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Isophthalic Acid Bis(2-ethylhexyl) Ester

Formula: C₂₄H₃₈O₄
Boiling Pt: 211 °C
Density: 0.98 (20 °C)
Flash Pt: 233 °C
MDL Number: MFCD00152284
CAS Number: 137-89-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ea2c11daf63bfa9ae4942f5f560c21597af55a5&vid=0ea2c11daf63bfa9ae4942f5f560c21597af55a5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR