4'-(Benzyloxy)benzylidene-4-fluoroaniline ≥98.0% (by GC)
Supplier: TCI America
B4301-25G
B4301-5G
TCB4301-25GEA
317.42
CAD
TCB4301-25G
TCB4301-5G
4'-(Benzyloxy)benzylidene-4-fluoroaniline ≥98.0% (by GC)
4'-(Benzyloxy)benzylidene-4-fluoroaniline
CAS Number: 70627-52-0
MDL Number: MFCD00017951
Molecular Formula: C20H16FNO
Molecular Weight: 305.35
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 136
Formula:
C₂₀H₁₆FNO Melting Pt: 136 °C |
MDL Number:
MFCD00017951 CAS Number: 70627-52-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...