N-(4-Bromophenyl)-N-phenyl-1-naphthylamine ≥98.0% (by GC)

Supplier: TCI America

B4299-1G B4299-200MG
TCB4299-1GEA 123.24 CAD
TCB4299-1G TCB4299-200MG
N-(4-Bromophenyl)-N-phenyl-1-naphthylamine ≥98.0% (by GC)
N-(4-Bromophenyl)-N-phenyl-1-naphthylamine

CAS Number: 138310-84-6
MDL Number: MFCD11046471
Molecular Formula: C22H16BrN
Molecular Weight: 374.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 93


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 1-[N-(4-Bromophenyl)anilino]naphthalene

Formula: C₂₂H₁₆BrN
Melting Pt: 93 °C
MDL Number: MFCD11046471
CAS Number: 138310-84-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7fe3ea07cc5e5e9b7732aaf7f97fa99ee8ec2cdd&vid=7fe3ea07cc5e5e9b7732aaf7f97fa99ee8ec2cdd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR