1-(2-Bromoethoxy)-4-chlorobenzene ≥98.0%

Supplier: TCI America

Synonyms: 4-Chloro-ω-bromophenetol, 4-Chlorophenyl 2-bromoethyl ether, 4-Chloro-β-bromophenetol

B4298-25G B4298-5G
TCB4298-25GEA 496.12 CAD
TCB4298-25G TCB4298-5G
1-(2-Bromoethoxy)-4-chlorobenzene ≥98.0%
1-(2-Bromoethoxy)-4-chlorobenzene

CAS Number: 2033-76-3
MDL Number: MFCD00000617
Molecular Formula: C8H8BrClO
Molecular Weight: 235.51
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 87
Melting point (°C): 42
Flash Point (°C): 113


Other Product Names / Synonyms: 2-Bromoethyl 4-Chlorophenyl Ether, beta-Bromo-4-chlorophenetole

Formula: C₈H₈BrClO
Boiling Pt: 87 °C
Melting Pt: 42 °C
Flash Pt: 113 °C
MDL Number: MFCD00000617
CAS Number: 2033-76-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f1af1e44de6be3267fc61740364a9194abb8e49&vid=9f1af1e44de6be3267fc61740364a9194abb8e49&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR