2,2'-Bipyrazine ≥97.0%

Supplier: TCI America

Synonyms: 2,2'-Bipyrazinyl

B4297-100MG B4297-500MG
TCB4297-100MGEA 176.96 CAD
TCB4297-100MG 76219-384
2,2'-Bipyrazine ≥97.0%
2,2'-Bipyrazine

CAS Number: 10199-00-5
MDL Number: MFCD00010288
Molecular Formula: C8H6N4
Molecular Weight: 158.16
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 188
Lambda max.: 320 nm (Hexane)


Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Metal Organic Framework (MOF) and Porous Coordination Polymer (PCP) Research Reagents
Other Product Names / Synonyms: 2,2'-Bipyrazinyl

Formula: C₈H₆N₄
MW: 158.16 g/mol
Melting Pt: 188 °C
MDL Number: MFCD00010288
CAS Number: 10199-00-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bd530dc244ec23b3ac8ac87f049c9339a808918&vid=1bd530dc244ec23b3ac8ac87f049c9339a808918&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR