(S)-2-((tert-Butoxycarbonyl)amino)-3-(pyridin-2-yl)propanoic acid ≥98.0%

Supplier: TCI America

Synonyms: N-Boc-3-(2-pyridyl)-L-alanine, Boc-2-Pal-OH

B4293-1G B4293-5G
TCB4293-1GEA 200.66 CAD
TCB4293-1G TCB4293-5G
(S)-2-((tert-Butoxycarbonyl)amino)-3-(pyridin-2-yl)propanoic acid ≥98.0%
(S)-2-((tert-Butoxycarbonyl)amino)-3-(pyridin-2-yl)propanoic acid

CAS Number: 71239-85-5
MDL Number: MFCD00191190
Molecular Formula: C13H18N2O4
Molecular Weight: 266.30
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 141
Specific rotation [a]20/D: -16 deg (C=1, EtOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-3-(2-pyridyl)-L-alanine, Boc-3-(2-pyridyl)-Ala-OH

Formula: C13H18N2O4
MW: 266.3 g/mol
Melting Pt: 141 °C
MDL Number: MFCD00191190
CAS Number: 71239-85-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0e965a3f5eb04f323b767e6542939aeee1345d4&vid=e0e965a3f5eb04f323b767e6542939aeee1345d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR