N-(tert-Butoxycarbonyl)-7-aminoheptanoic acid ≥98.0%

Supplier: TCI America

Synonyms: Boc-7-Ah, 7-((tert-Butoxycarbonyl)amino)heptanoic acid, 7-(tert-Butoxycarbonylamino)enanthic Acid, 7-(Boc-amino)enanthic Acid, N-(tert-Butoxycarbonyl)-7-aminoenanthic Acid, 7-(tert-Butoxycarbonylamino)heptanoic Acid, N-Boc-7-aminoheptanoic Acid, 7-(Boc-amino)heptanoic Acid, N-Boc-7-aminoenanthic Acid

B4292-1G B4292-5G
TCB4292-1GEA 86.9 CAD
TCB4292-1G TCB4292-5G
N-(tert-Butoxycarbonyl)-7-aminoheptanoic acid ≥98.0%
N-(tert-Butoxycarbonyl)-7-aminoheptanoic acid

CAS Number: 60142-89-4
MDL Number: MFCD00063356
Molecular Formula: C12H23NO4
Molecular Weight: 245.32
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 57
Storage Temperature: 0-10°C


Product Categories: Bioscience, Amino Acids (N-Protected)

Formula: C₁₂H₂₃NO₄
MW: 245.32 g/mol
Melting Pt: 57 °C
MDL Number: MFCD00063356
CAS Number: 60142-89-4

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR