N-(tert-Butoxycarbonyl)-4-aminobutyric acid ≥98.0%

Supplier: TCI America

Synonyms: Boc-GABA-OH, N-Boc-γ-aminobutyric acid, 4-(Boc-amino)butyric Acid, 4-(tert-Butoxycarbonylamino)butyric Acid, N-Boc-4-aminobutyric Acid, N-BOC-gamma-Aminobutyric acid

B4291-25G B4291-5G
TCB4291-25GEA 290.89 CAD
TCB4291-25G TCB4291-5G
N-(tert-Butoxycarbonyl)-4-aminobutyric acid ≥98.0%
N-(tert-Butoxycarbonyl)-4-aminobutyric acid

CAS Number: 57294-38-9
MDL Number: MFCD00037313
Molecular Formula: C9H17NO4
Molecular Weight: 203.24
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 82
Melting point (°C): 58


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Boc-4-aminobutyric Acid, 4-(tert-Butoxycarbonylamino)butyric Acid, 4-(Boc-amino)butyric Acid, Boc-GABA-OH

Formula: C₉H₁₇NO₄
MW: 203.24 g/mol
Boiling Pt: 82 °C
Melting Pt: 58 °C
MDL Number: MFCD00037313
CAS Number: 57294-38-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b44978372b6e07ceeec6a64b1658b85bdec3fd9&vid=3b44978372b6e07ceeec6a64b1658b85bdec3fd9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR