tert-Butyl-3-iodoazetidine-1-carboxylate ≥95.0% (by GC)

Supplier: TCI America

B4288-1G B4288-5G
TCB4288-1GEA 56.88 CAD
TCB4288-1G TCB4288-5G
tert-Butyl-3-iodoazetidine-1-carboxylate ≥95.0% (by GC)
tert-Butyl-3-iodoazetidine-1-carboxylate

CAS Number: 254454-54-1
MDL Number: MFCD09752821
Molecular Formula: C8H14INO2
Molecular Weight: 283.11
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.51


Other Product Names / Synonyms: 1-Boc-3-iodoazetidine, tert-Butyl 3-Iodoazetidine-1-carboxylate, 3-Iodoazetidine-1-carboxylic Acid tert-Butyl Ester

Formula: C₈H₁₄INO₂
Density: 1.51 (20 °C)
MDL Number: MFCD09752821
CAS Number: 254454-54-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07a5a1ba2ac6cfadfa0d273020e4cc732d5c10a8&vid=07a5a1ba2ac6cfadfa0d273020e4cc732d5c10a8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR