N-(tert-Butoxycarbonyl)-L-methioninol ≥95.0%

Supplier: TCI America

Synonyms: (S)-2-(tert-Butoxycarbonyl)amino-4-methylthio-1-butanol, Boc-Methioninol, N-Boc-L-methioninol, (S)-2-(Boc-amino)-4-methylthio-1-butanol, (S)-tert-Butyl (1-hydroxy-4-(methylthio)butan-2-yl)carbamate

B4283-1G B4283-5G
TCB4283-1GEA 57.83 CAD
TCB4283-1G TCB4283-5G
N-(tert-Butoxycarbonyl)-L-methioninol ≥95.0%
N-(tert-Butoxycarbonyl)-L-methioninol

CAS Number: 51372-93-1
MDL Number: MFCD00235939
Molecular Formula: C10H21NO3S
Molecular Weight: 235.34
Purity/Analysis Method: >95.0% (GC)
Form: Crystalline Powder
Melting point (°C): 50
Specific rotation [a]20/D: -20 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: N-Boc-L-methioninol, (S)-2-(tert-Butoxycarbonyl)amino-4-methylthio-1-butanol, (S)-2-(Boc-amino)-4-methylthio-1-butanol

Formula: C₁₀H₂₁NO₃S
MW: 235.34 g/mol
Melting Pt: 50 °C
MDL Number: MFCD00235939
CAS Number: 51372-93-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85b091cc31c580b4c0803986f84b532ecb8f57e5&vid=85b091cc31c580b4c0803986f84b532ecb8f57e5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR