3-(Boc-amino)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
B4279-1G
B4279-5G
TCB4279-1GEA
93.22
CAD
TCB4279-1G
TCB4279-5G
3-(Boc-amino)phenylboronic acid (contains varying amounts of Anhydride)
3-(Boc-amino)phenylboronic acid
CAS Number: 380430-68-2
MDL Number: MFCD03411945
Molecular Formula: C11H16BNO4
Molecular Weight: 237.06
Form: Crystal
Color: White
Melting point (°C): 168
Other Product Names / Synonyms: 3-[(tert-Butoxycarbonyl)amino]benzeneboronic Acid, 3-(Boc-amino)phenylboronic Acid, 3-(Boc-amino)benzeneboronic Acid
Formula:
C₁₁H₁₆BNO₄ Melting Pt: 168 °C |
MDL Number:
MFCD03411945 CAS Number: 380430-68-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...