6,7-Bis(2-methoxyethoxy)-3H-quinazolin-4-one ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B4270-1G B4270-5G
TCB4270-1GEA 76 CAD
TCB4270-1G TCB4270-5G
6,7-Bis(2-methoxyethoxy)-3H-quinazolin-4-one ≥98.0% (by HPLC, titration analysis)
6,7-Bis(2-methoxyethoxy)-3H-quinazolin-4-one

CAS Number: 179688-29-0
MDL Number: MFCD02678087
Molecular Formula: C14H18N2O5
Molecular Weight: 294.31
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 192


Other Product Names / Synonyms: 6,7-Bis(2-methoxyethoxy)-4(3H)-quinazolinone, 6,7-Bis(2-methoxyethoxy)-3,4-dihydroquinazolin-4-one, 6,7-Bis(2-methoxyethoxy)-4-hydroxyquinazoline, 6,7-Bis(2-methoxyethoxy)-4-quinazolinol

Formula: C₁₄H₁₈N₂O₅
Melting Pt: 192 °C
MDL Number: MFCD02678087
CAS Number: 179688-29-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b7906d35e7e9dce2951a270dee82cc214060fa7&vid=2b7906d35e7e9dce2951a270dee82cc214060fa7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR