9,9-Bis(4-glycidyloxyphenyl)fluorene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4,4'-(9-Fluorenylidene)bis(1,2-epoxy-3-phenoxypropane), 2,2'-((((9H-Fluorene-9,9-diyl)bis(4,1-phenylene))bis(oxy))bis(methylene))bis(oxirane)

B4269-25G B4269-5G
TCB4269-25GEA 284.24 CAD
TCB4269-25G TCB4269-5G
9,9-Bis(4-glycidyloxyphenyl)fluorene ≥98.0% (by GC)
9,9-Bis(4-glycidyloxyphenyl)fluorene

CAS Number: 47758-37-2
MDL Number: MFCD02171148
Molecular Formula: C31H26O4
Molecular Weight: 462.55
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 159


Product Categories: Materials Chemistry, Monomers, Photopolymer Reagents
Other Product Names / Synonyms: 4,4'-(9-Fluorenylidene)bis(1,2-epoxy-3-phenoxypropane)

Formula: C₃₁H₂₆O₄
MW: 462.55 g/mol
Melting Pt: 159 °C
MDL Number: MFCD02171148
CAS Number: 47758-37-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f388a99fae541d9ecef4de223459a17917274399&vid=f388a99fae541d9ecef4de223459a17917274399&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR