N,N'-Bis(2,6-diisopropylphenyl)-3,4,9,10-perylenetetracarboxylic diimide ≥98.0%

Supplier: TCI America

Synonyms: Perylene Orange

B4268-1G B4268-5G
TCB4268-1GEA 641.32 CAD
TCB4268-1G TCB4268-5G
N,N'-Bis(2,6-diisopropylphenyl)-3,4,9,10-perylenetetracarboxylic diimide ≥98.0%
N,N'-Bis(2,6-diisopropylphenyl)-3,4,9,10-perylenetetracarboxylic diimide

CAS Number: 82953-57-9
Molecular Formula: C48H42N2O4
Molecular Weight: 710.87
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Lambda max.: 527 nm (Toluene)


Product Categories: Materials Chemistry, Organic Transistor Materials, Solar Cell Research Reagents, Perylene Dyes (Functional Organic Dyes)
Other Product Names / Synonyms: Perylene Orange

Formula: C₄₈H₄₂N₂O₄
MW: 710.87 g/mol
CAS Number: 82953-57-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fcadbfa67ee4f07afbfafb89136961a2ca185f54&vid=fcadbfa67ee4f07afbfafb89136961a2ca185f54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR