tert-Butyl-5-norbornene-2-carboxylate (endo and exo mixture) ≥95.0%
Supplier: TCI America
Synonyms:
5-Norbornene-2-carboxylic Acid tert-Butyl Ester, tert-Butyl Bicyclo[2.2.1]hept-5-ene-2-carboxylate, Bicyclo[2.2.1]hept-5-ene-2-carboxylic Acid tert-Butyl Ester, tert-Butyl Bicyclo[2.2.1]-5-heptene-2-carboxylate
CAS Number: 154970-45-3
MDL Number: MFCD18083704
Molecular Formula: C12H18O2
Molecular Weight: 194.27
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 103
Specific Gravity (20/20): 0.98
Other Product Names / Synonyms: 5-Norbornene-2-carboxylic Acid tert-Butyl Ester, tert-Butyl Bicyclo[2.2.1]hept-5-ene-2-carboxylate, Bicyclo[2.2.1]hept-5-ene-2-carboxylic Acid tert-Butyl Ester
Formula:
C₁₂H₁₈O₂ MW: 194.27 g/mol Boiling Pt: 103 °C Density: 0.98 (20 °C) |
MDL Number:
MFCD18083704 CAS Number: 154970-45-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...