Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride ≥98.0% (by GC, titration analysis), purified by sublimation

Supplier: TCI America

B4261-25G B4261-5G
TCB4261-25GEA 395 CAD
TCB4261-25G TCB4261-5G
Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride ≥98.0% (by GC, titration analysis), purified by sublimation
Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride

CAS Number: 1719-83-1
MDL Number: MFCD00082228
Molecular Formula: C12H8O6
Molecular Weight: 248.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 382

Formula: C₁₂H₈O₆
Melting Pt: 382 °C
MDL Number: MFCD00082228
CAS Number: 1719-83-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd99aacfbf5e4dcfaca2b09409beec8843df7e57&vid=2b1c1c464921cd47240ced1f849fa15c668bc99d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR