3,3',4,4'-Benzophenonetetracarboxylic dianhydride ≥98.0% (by GC), purified by sublimation

Supplier: TCI America

B4260-25G B4260-5G
TCB4260-25GEA 423.12 CAD
TCB4260-25G TCB4260-5G
3,3',4,4'-Benzophenonetetracarboxylic dianhydride ≥98.0% (by GC), purified by sublimation
3,3',4,4'-Benzophenonetetracarboxylic dianhydride

CAS Number: 2421-28-5
MDL Number: MFCD00005923
Molecular Formula: C17H6O7
Molecular Weight: 322.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 227
Flash Point (°C): 324
Lambda max.: 580 nm (CH3CN)


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials, Benzophenones (for High-Performance Polymer Research), Highly Purified Reagents
Other Product Names / Synonyms: 4,4'-Carbonyldiphthalic Anhydride

Formula: C₁₇H₆O₇
Melting Pt: 227 °C
Flash Pt: 324 °C
MDL Number: MFCD00005923
CAS Number: 2421-28-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba51e47cbca5f7278926e6859b6c2121f8c61948&vid=f94e7d0cfdf0d8bcc7ee5105c37089dcc113f3cd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR