N-(tert-Butoxycarbonyl)-L-phenylalanine methyl ester ≥98.0%

Supplier: TCI America

Synonyms: Methyl (tert-butoxycarbonyl)-L-phenylalaninate, N-Boc-L-phenylalanine Methyl Ester, Boc-Phe-OMe

B4259-25G B4259-5G
TCB4259-25GEA 150.1 CAD
TCB4259-25G TCB4259-5G
N-(tert-Butoxycarbonyl)-L-phenylalanine methyl ester ≥98.0%
N-(tert-Butoxycarbonyl)-L-phenylalanine methyl ester

CAS Number: 51987-73-6
MDL Number: MFCD00076977
Molecular Formula: C15H21NO4
Molecular Weight: 279.34
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 39
Flash Point (°C): 113
Specific rotation [a]20/D: -3 deg (C=2, MeOH)


Product Categories: Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: N-Boc-L-phenylalanine Methyl Ester, Boc-Phe-OMe

Formula: C₁₅H₂₁NO₄
MW: 279.34 g/mol
Melting Pt: 39 °C
Flash Pt: 113 °C
MDL Number: MFCD00076977
CAS Number: 51987-73-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d40e34a7bf50e67c555e55bde2b922a27a052f4&vid=4d40e34a7bf50e67c555e55bde2b922a27a052f4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR