1,3-Bis[5-(4-tert-butylphenyl)-2-[1,3,4]oxadiazolyl]benzene ≥97.0% (by HPLC)
Supplier: TCI America
B4252-1G
B4252-200MG
TCB4252-1GEA
417.12
CAD
TCB4252-1G
TCB4252-200MG
1,3-Bis[5-(4-tert-butylphenyl)-2-[1,3,4]oxadiazolyl]benzene ≥97.0% (by HPLC)
1,3-Bis[5-(4-tert-butylphenyl)-2-[1,3,4]oxadiazolyl]benzene
CAS Number: 138372-67-5
MDL Number: MFCD03093214
Molecular Formula: C30H30N4O2
Molecular Weight: 478.60
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Melting point (°C): 241
Lambda max.: 292 nm (CH2Cl2)
Product Categories: Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials
Other Product Names / Synonyms: 1,3-Bis[5-(4-tert-butylphenyl)-1,3,4-oxadiazol-2-yl]benzene, OXD-7
Formula:
C₃₀H₃₀N₄O₂ Melting Pt: 241 °C |
MDL Number:
MFCD03093214 CAS Number: 138372-67-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...