tert-Butyl (1-(hydroxymethyl)cyclopropyl)carbamate ≥97.0%

Supplier: TCI America

Synonyms: [1-(Hydroxymethyl)cyclopropyl]carbamic acid tert-Butyl ester, [1-(Boc-amino)cyclopropyl]methanol, tert-Butyl [1-(Hydroxymethyl)cyclopropyl]carbamate, [1-(tert-Butoxycarbonylamino)cyclopropyl]methanol

B4251-1G B4251-200MG
TCB4251-1GEA 196.71 CAD
TCB4251-1G TCB4251-200MG
tert-Butyl (1-(hydroxymethyl)cyclopropyl)carbamate ≥97.0%
tert-Butyl (1-(hydroxymethyl)cyclopropyl)carbamate

CAS Number: 107017-73-2
MDL Number: MFCD09749954
Molecular Formula: C9H17NO3
Molecular Weight: 187.24
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 83


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: [1-(Boc-amino)cyclopropyl]methanol, tert-Butyl [1-(Hydroxymethyl)cyclopropyl]carbamate, [1-(Hydroxymethyl)cyclopropyl]carbamic Acid tert-Butyl Ester

Formula: C₉H₁₇NO₃
MW: 187.2362 g/mol
Melting Pt: 83 °C
MDL Number: MFCD09749954
CAS Number: 107017-73-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fcf35a7746391eece94e8062f140aa576bd1b400&vid=fcf35a7746391eece94e8062f140aa576bd1b400&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR