tert-Butyl-3-(hydroxymethyl)pyrrolidine-1-carboxylate ≥95.0%
Supplier: TCI America
Synonyms:
tert-Butyl 3-(hydroxymethyl)pyrrolidine-1-carboxylate, 1-BOC-3-Pyrrolidinemethanol, 1-Boc-3-(hydroxymethyl)pyrrolidine
CAS Number: 114214-69-6
MDL Number: MFCD02179040
Molecular Formula: C10H19NO3
Molecular Weight: 201.27
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 291
Specific Gravity (20/20): 1.06
Other Product Names / Synonyms: 1-Boc-3-pyrrolidinemethanol, tert-Butyl 3-(Hydroxymethyl)pyrrolidine-1-carboxylate, 3-(Hydroxymethyl)pyrrolidine-1-carboxylic Acid tert-Butyl Ester
Formula:
C₁₀H₁₉NO₃ MW: 201.27 g/mol Boiling Pt: 291 °C Density: 1.06 (20 °C) |
MDL Number:
MFCD02179040 CAS Number: 114214-69-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...