4-Bromo-DL-phenylalanine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B4245-1G B4245-5G
TCB4245-1GEA 66.36 CAD
TCB4245-1G TCB4245-5G
4-Bromo-DL-phenylalanine ≥98.0% (by HPLC, titration analysis)
4-Bromo-DL-phenylalanine

CAS Number: 14091-15-7
MDL Number: MFCD00002599
Molecular Formula: C9H10BrNO2
Molecular Weight: 244.09
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 258
Storage Temperature: 0-10°C


Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: 2-Amino-3-(4-bromophenyl)propionic Acid, H-DL-Phe(4-Br)-OH

Formula: C₉H₁₀BrNO₂
Melting Pt: 258 °C
MDL Number: MFCD00002599
CAS Number: 14091-15-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e713c28a03fcd40702c8d5e27021fd803052b0ab&vid=e713c28a03fcd40702c8d5e27021fd803052b0ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR