1-Bromo-5-chloro-2-iodobenzene ≥96.0% (by GC)

Supplier: TCI America

B4243-25G B4243-5G
TCB4243-25GEA 195.92 CAD
TCB4243-25G TCB4243-5G
1-Bromo-5-chloro-2-iodobenzene ≥96.0% (by GC)
1-Bromo-5-chloro-2-iodobenzene

CAS Number: 31928-44-6
MDL Number: MFCD00672944
Molecular Formula: C6H3BrClI
Molecular Weight: 317.35
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Boiling point (°C): 111
Melting point (°C): 33

Formula: C₆H₃BrClI
Boiling Pt: 111 °C
Melting Pt: 33 °C
MDL Number: MFCD00672944
CAS Number: 31928-44-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a63382c7ce1de254f911fba4850ef4acfc567677&vid=a63382c7ce1de254f911fba4850ef4acfc567677&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR