2,5-Bis[(trimethylsilyl)ethynyl]thiophene ≥96.0% (by GC)

Supplier: TCI America

B4240-1G B4240-5G
TCB4240-1GEA 261.17 CAD
TCB4240-1G TCB4240-5G
2,5-Bis[(trimethylsilyl)ethynyl]thiophene ≥96.0% (by GC)
2,5-Bis[(trimethylsilyl)ethynyl]thiophene

CAS Number: 79109-69-6
MDL Number: MFCD18207392
Molecular Formula: C14H20SSi2
Molecular Weight: 276.54
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 82
Lambda max.: 331 nm (CHCl3)

Formula: C₁₄H₂₀SSi₂
Melting Pt: 82 °C
MDL Number: MFCD18207392
CAS Number: 79109-69-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb480c8a50adf6bf0e5cc73154abaec334472b80&vid=bb480c8a50adf6bf0e5cc73154abaec334472b80&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR