9-Bromo-9,10-dihydro-9,10-ethenoanthracene ≥98.0% (by GC)

Supplier: TCI America

B4239-1G B4239-5G
TCB4239-1GEA 382.36 CAD
TCB4239-1G TCB4239-5G
9-Bromo-9,10-dihydro-9,10-ethenoanthracene ≥98.0% (by GC)
9-Bromo-9,10-dihydro-9,10-ethenoanthracene

CAS Number: 126690-96-8
Molecular Formula: C16H11Br
Molecular Weight: 283.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 140


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 9-Bromodibenzobarrelene

Formula: C₁₆H₁₁Br
Melting Pt: 140 °C
CAS Number: 126690-96-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0332a5daf36020968e5faee656a7cb0565decf25&vid=0332a5daf36020968e5faee656a7cb0565decf25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR