tert-Butyl (2-aminophenyl)carbamate ≥98.0%

Supplier: TCI America

Synonyms: N-(tert-Butoxycarbonyl)-1,2-phenylenediamine, tert-Butyl (2-Aminophenyl)carbamate, 2-(tert-Butoxycarbonylamino)aniline, (2-Aminophenyl)carbamic acid tert-Butyl ester, 2-(Boc-amino)aniline, N-Boc-1,2-phenylenediamine

B4238-1G B4238-5G
TCB4238-1GEA 125.61 CAD
TCB4238-1G TCB4238-5G
tert-Butyl (2-aminophenyl)carbamate ≥98.0%
tert-Butyl (2-aminophenyl)carbamate

CAS Number: 146651-75-4
MDL Number: MFCD02169707
Molecular Formula: C11H16N2O2
Molecular Weight: 208.26
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 116


Other Product Names / Synonyms: N-Boc-1,2-phenylenediamine, tert-Butyl (2-Aminophenyl)carbamate, (2-Aminophenyl)carbamic Acid tert-Butyl Ester, 2-(tert-Butoxycarbonylamino)aniline, 2-(Boc-amino)aniline

Formula: C₁₁H₁₆N₂O₂
MW: 208.2600 g/mol
Melting Pt: 116 °C
MDL Number: MFCD02169707
CAS Number: 146651-75-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae5c0f4280492eebf132837da640c4c479b464d7&vid=ae5c0f4280492eebf132837da640c4c479b464d7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR