N-Boc-m-phenylenediamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B4222-1G B4222-5G
TCB4222-1GEA 94.8 CAD
TCB4222-1G TCB4222-5G
N-Boc-m-phenylenediamine ≥98.0% (by GC, titration analysis)
N-Boc-m-phenylenediamine

CAS Number: 68621-88-5
MDL Number: MFCD06657915
Molecular Formula: C11H16N2O2
Molecular Weight: 208.26
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal


Other Product Names / Synonyms: N-Boc-1,3-phenylenediamine, tert-Butyl (3-Aminophenyl)carbamate, (3-Aminophenyl)carbamic Acid tert-Butyl Ester, 3-(tert-Butoxycarbonylamino)aniline, 3-(Boc-amino)aniline

Formula: C₁₁H₁₆N₂O₂
MDL Number: MFCD06657915
CAS Number: 68621-88-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c39679c757bad23e5e7e64295f9dfaea3d03e3f&vid=7c39679c757bad23e5e7e64295f9dfaea3d03e3f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR