2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene ≥99.0% (by HPLC, total nitrogen), purified by sublimation
Supplier: TCI America
B4221-1G
B4221-5G
TCB4221-1GEA
201.61
CAD
TCB4221-1G
TCB4221-5G
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene ≥99.0% (by HPLC, total nitrogen), purified by sublimation
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene
CAS Number: 7128-64-5
MDL Number: MFCD00005774
Molecular Formula: C26H26N2O2S
Molecular Weight: 430.57
Purity/Analysis Method: >99.0% (HPLC,N)
Form: Crystal
Melting point (°C): 202
Lambda max.: 373 nm (EtOH)
Formula:
C₂₆H₂₆N₂O₂S Melting Pt: 202 °C |
MDL Number:
MFCD00005774 CAS Number: 7128-64-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...