Bromobimane (mBBr) ≥98.0% (by HPLC)

Supplier: TCI America

B4220-100MG B4220-20MG
TCB4220-100MGEA 1096.14 CAD
TCB4220-100MG TCB4220-20MG
Bromobimane (mBBr) ≥98.0% (by HPLC)
Bromobimane (mBBr)

CAS Number: 71418-44-5
MDL Number: MFCD00036951
Molecular Formula: C10H11BrN2O2
Molecular Weight: 271.11
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 162
Lambda max.: 395 nm (H2O)Storage Temperature: <0°C


Product Categories: Bioscience, Fluorescent Labeling Dyes, Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: Monobromobimane, 3-(Bromomethyl)-2,5,6-trimethylpyrazolo[1,2-a]pyrazole-1,7-dione

Formula: C₁₀H₁₁BrN₂O₂
Melting Pt: 162 °C
MDL Number: MFCD00036951
CAS Number: 71418-44-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57af284b2aa088e37ab57807c0f467dc56cce331&vid=57af284b2aa088e37ab57807c0f467dc56cce331&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR